Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172457.2 | NP_766045.2 | 2 - 3 | 524 | 71 | |
GenBank mRNA | AK030481.1 | - | 2 - 3 | 560 | 71 | |
AK170394.1 | - | 2 - 3 | 535 | 71 | ||
AK171197.1 | - | 548 | 71 | |||
BC051170.1 | - | 1 - 2 | 513 | 71 | ||
BC058238.1 | - | 2 - 3 | 524 | 6307543 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |