Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163165.1 | NP_001156637.1 | 6 - 7 | 693 | 69 | |
NM_001163166.1 | NP_001156638.1 | 6 - 7 | 590 | 69 | ||
NM_010440.3 | NP_034570.1 | 5 - 6 | 777 | 69 | ||
GenBank mRNA | AF146224.1 | - | 5 - 6 | 760 | 69 | |
AK153816.1 | - | 6 - 7 | 591 | 69 | ||
AK155652.1 | - | 6 - 7 | 595 | 69 | ||
AK155890.1 | - | 5 - 6 | 653 | 69 | ||
AK189436.1 | - | 2 - 3 | 170 | 69 | ||
AL355735.1 | - | 2 - 3 | 152 | 69 | ||
BC003924.1 | - | 5 - 6 | 665 | 3485302 | 69 | |
BC011334.1 | - | 6 - 7 | 626 | 4012515 | 69 | |
BC023436.1 | - | 3 - 4 | 253 | 69 | ||
BC071217.1 | - | 3 - 4 | 1654 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |