Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019412.2 | NP_062285.1 | 2 - 3 | 90 | 74 | |
NM_198048.2 | NP_932165.2 | 2 - 3 | 90 | 74 | ||
GenBank mRNA | AJ222968.1 | - | 2 - 3 | 90 | 74 | |
AJ222969.1 | - | 2 - 3 | 90 | 74 | ||
CJ045218.1 | - | 2 - 3 | 93 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |