Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290469.1 | NP_001277398.1 | 7 - 8 | 899 | 78 | |
GenBank mRNA | AK053751.1 | - | 7 - 8 | 871 | 78 | |
BC020177.1 | - | 7 - 8 | 857 | 4506376 | 78 | |
BC034645.1 | - | 7 - 8 | 855 | 5357731 | 78 | |
BC037206.1 | - | 7 - 8 | 854 | 5362423 | 78 | |
BC042894.1 | - | 7 - 8 | 899 | 5364812 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |