Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pofut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030262.3 | NP_084538.2 | 2 - 3 | 408 | 67 | |
GenBank mRNA | AF455270.1 | - | 2 - 3 | 398 | 67 | |
AK132373.1 | - | 2 - 3 | 405 | 67 | ||
AK148244.1 | - | 2 - 3 | 407 | 67 | ||
BC018194.1 | - | 2 - 3 | 406 | 67 | ||
BC147077.1 | - | 2 - 3 | 475 | 9056064 | 67 | |
BC147078.1 | - | 2 - 3 | 475 | 9056063 | 67 | |
CO814561.1 | - | 408 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |