Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010925.2 | NP_035055.2 | 1 - 2 | 223 | 101 | |
GenBank mRNA | AK077540.1 | - | 1 - 2 | 148 | 101 | |
BC065993.1 | - | 1 - 2 | 185 | 5684672 | 101 | |
BC065994.1 | - | 1 - 2 | 143 | 5685269 | 101 | |
BY106122.1 | - | 1 - 2 | 223 | 101 | ||
U79773.1 | - | 1 - 2 | 175 | 101 | ||
U79774.1 | - | 1 - 2 | 175 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |