Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011820.4 | NP_035950.2 | 4 - 5 | 924 | 64 | |
GenBank mRNA | AF077765.1 | - | 4 - 5 | 909 | 64 | |
AK049084.1 | - | 4 - 5 | 926 | 64 | ||
AK052183.1 | - | 4 - 5 | 925 | 64 | ||
AK170326.1 | - | 4 - 5 | 788 | 64 | ||
BC113775.1 | - | 4 - 5 | 648 | 40059928 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |