Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
NM_001017407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
NM_001017407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
NM_001017409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
NM_001017409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
NM_023064.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
NM_023064.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003799.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017441.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_001017441.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_023064.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
NM_023064.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003801.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017441.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_001017441.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_023064.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
NM_023064.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003802.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
NM_001017419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
NM_001017419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
NM_001017433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
NM_001017433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
NM_001017441.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
NM_001017441.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003797.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
NM_001017407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
NM_001017407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
NM_001017409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
NM_001017409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
NM_023064.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
NM_023064.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003798.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017441.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_001017441.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_023064.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
NM_023064.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003805.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003806.mm.1 |
NM_001017409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003806.mm.1 |
NM_001017409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003806.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
NM_001017419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
NM_001017419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
NM_001017433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
NM_001017433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
NM_001017441.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
NM_001017441.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003795.mm.1 |
TC1000000560.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
NM_001017419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
NM_001017419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
NM_001017433.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
NM_001017433.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
NM_001017441.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
NM_001017441.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000003796.mm.1 |
TC1000000560.mm.1 |
MTA TC |