Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008793.2 | NP_032819.1 | 13 - 14 | 1714 | 64 | |
GenBank mRNA | AK161504.1 | - | 11 - 12 | 1973 | 64 | |
BC052535.1 | - | 1 - 2 | 412 | 64 | ||
D01093.1 | - | 13 - 14 | 1714 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |