Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195268.1 | NP_001182197.1 | 4 - 5 | 453 | 59 | |
NR_036582.1 | 6 - 7 | 789 | 59 | |||
GenBank mRNA | AK137009.1 | - | 6 - 7 | 789 | 59 | |
BC052075.1 | - | 5 - 6 | 507 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |