Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Theg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199226.1 | NP_954696.1 | 2 - 3 | 475 | 66 | |
GenBank mRNA | BC050970.1 | - | 2 - 3 | 476 | 66 | |
BC061243.1 | - | 2 - 3 | 421 | 6775002 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |