Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164248.1 | NP_001157720.1 | 2 - 3 | 323 | 66 | |
NM_001164250.1 | NP_001157722.1 | 2 - 3 | 323 | 66 | ||
NM_001164251.1 | NP_001157723.1 | 2 - 3 | 323 | 66 | ||
NM_001164255.1 | NP_001157727.1 | 2 - 3 | 323 | 66 | ||
NM_024427.4 | NP_077745.2 | 2 - 3 | 323 | 66 | ||
GenBank mRNA | AK002271.1 | - | 2 - 3 | 324 | 66 | |
AK003175.1 | - | 2 - 3 | 324 | 66 | ||
AK032942.1 | - | 2 - 3 | 372 | 66 | ||
AK077713.1 | - | 2 - 3 | 324 | 66 | ||
AK168640.1 | - | 2 - 3 | 320 | 66 | ||
AK168783.1 | - | 2 - 3 | 320 | 66 | ||
AK169188.1 | - | 2 - 3 | 324 | 66 | ||
BC026720.1 | - | 2 - 3 | 297 | 66 | ||
BC132035.1 | - | 2 - 3 | 282 | 40130312 | 66 | |
BC132037.1 | - | 2 - 3 | 282 | 40130314 | 66 | |
M22479.1 | - | 2 - 3 | 296 | 66 | ||
X64831.1 | - | 2 - 3 | 278 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |