Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027261.3 | NP_081537.1 | 2 - 3 | 164 | 85 | |
NM_029248.2 | NP_083524.2 | 2 - 3 | 164 | 85 | ||
NR_028401.1 | 2 - 3 | 164 | 85 | |||
GenBank mRNA | AK012662.1 | - | 2 - 3 | 116 | 85 | |
AK014920.1 | - | 2 - 3 | 116 | 85 | ||
AK016109.1 | - | 2 - 3 | 118 | 85 | ||
AK050165.1 | - | 2 - 3 | 120 | 85 | ||
AK161656.1 | - | 2 - 3 | 119 | 85 | ||
AK166633.1 | - | 2 - 3 | 119 | 85 | ||
BC029906.1 | - | 1 - 2 | 619 | 85 | ||
BC056964.1 | - | 1 - 2 | 102 | 85 | ||
BC110660.1 | - | 1 - 2 | 106 | 3158156 | 85 | |
BC117068.1 | - | 1 - 2 | 96 | 40126387 | 85 | |
BC117070.1 | - | 1 - 2 | 96 | 40126389 | 85 | |
BU850945.1 | - | 2 - 3 | 168 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |