Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028401.1 | 6 - 7 | 945 | 113 | ||
GenBank mRNA | AK012656.1 | - | 4 - 5 | 794 | 113 | |
AK014920.1 | - | 6 - 7 | 897 | 113 | ||
AK020036.1 | - | 5 - 6 | 857 | 113 | ||
AK050165.1 | - | 6 - 7 | 901 | 113 | ||
AK166329.1 | - | 6 - 7 | 900 | 113 | ||
AK166633.1 | - | 5 - 6 | 2015 | 113 | ||
BC110660.1 | - | 5 - 6 | 887 | 3158156 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |