Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039520.2 | NP_001034609.1 | 16 - 17 | 1966 | 65 | |
NM_001253893.1 | NP_001240822.1 | 16 - 17 | 1966 | 65 | ||
NM_001253894.1 | NP_001240823.1 | 16 - 17 | 1966 | 65 | ||
NM_007871.2 | NP_031897.2 | 15 - 16 | 1954 | 65 | ||
GenBank mRNA | AK005012.2 | - | 16 - 17 | 1942 | 65 | |
AK150046.1 | - | 16 - 17 | 1929 | 65 | ||
AK154375.1 | - | 14 - 15 | 1618 | 65 | ||
AK157844.1 | - | 16 - 17 | 1935 | 65 | ||
AK170574.1 | - | 16 - 17 | 1942 | 65 | ||
AK171566.1 | - | 16 - 17 | 1930 | 65 | ||
AK172231.1 | - | 16 - 17 | 1947 | 65 | ||
BC010233.1 | - | 7 - 8 | 682 | 3499181 | 65 | |
L31398.1 | - | 15 - 16 | 1894 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |