Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007503.3 | NP_031529.2 | 4 - 5 | 243 | 86 | |
NM_052823.2 | NP_439888.1 | 4 - 5 | 316 | 86 | ||
NR_028499.1 | 4 - 5 | 448 | 86 | |||
GenBank mRNA | AA396010.1 | - | 4 - 5 | 264 | 86 | |
AI316264.1 | - | 4 - 5 | 227 | 86 | ||
AK002215.1 | - | 4 - 5 | 231 | 86 | ||
AK002228.1 | - | 4 - 5 | 244 | 86 | ||
AK007105.1 | - | 2 - 3 | 118 | 86 | ||
AY626243.1 | - | 4 - 5 | 365 | 86 | ||
AY626244.1 | - | 4 - 5 | 316 | 86 | ||
AY626245.1 | - | 4 - 5 | 401 | 86 | ||
BC024614.1 | - | 4 - 5 | 232 | 4222609 | 86 | |
BU756853.1 | - | 2 - 3 | 86 | |||
CB600439.1 | - | 4 - 5 | 276 | 86 | ||
CB600958.1 | - | 4 - 5 | 451 | 86 | ||
X70060.1 | - | 3 - 4 | 154 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |