Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176831.4 | NP_789801.1 | 4 - 5 | 389 | 93 | |
GenBank mRNA | AK036042.1 | - | 4 - 5 | 372 | 93 | |
AK038628.1 | - | 4 - 5 | 367 | 93 | ||
AK046751.1 | - | 1 - 2 | 223 | 93 | ||
AK078825.1 | - | 4 - 5 | 372 | 93 | ||
BC004779.1 | - | 4 - 5 | 382 | 93 | ||
BC052928.1 | - | 3 - 4 | 338 | 5255304 | 93 | |
BC089351.1 | - | 3 - 4 | 353 | 30603115 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |