Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 3 - 4 | 500 | 64 | |
GenBank mRNA | AK004783.1 | - | 3 - 4 | 484 | 64 | |
AK032030.1 | - | 1 - 2 | 121 | 64 | ||
AK154642.1 | - | 3 - 4 | 413 | 64 | ||
BC037093.1 | - | 3 - 4 | 521 | 64 | ||
BC151153.1 | - | 3 - 4 | 426 | 9088047 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |