Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 4 - 5 | 602 | 66 | |
GenBank mRNA | AK004783.1 | - | 4 - 5 | 586 | 66 | |
AK032030.1 | - | 2 - 3 | 223 | 66 | ||
AK154642.1 | - | 4 - 5 | 515 | 66 | ||
AK205345.1 | - | 1 - 2 | 51 | 66 | ||
BC037093.1 | - | 4 - 5 | 623 | 66 | ||
BC151153.1 | - | 4 - 5 | 528 | 9088047 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |