Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027895.1 | NP_082171.1 | 5 - 6 | 749 | 100 | |
GenBank mRNA | AK032030.1 | - | 3 - 4 | 370 | 100 | |
AK154642.1 | - | 5 - 6 | 662 | 100 | ||
BC037093.1 | - | 5 - 6 | 770 | 100 | ||
BC039958.1 | - | 1 - 2 | 111 | 100 | ||
BC151153.1 | - | 5 - 6 | 675 | 9088047 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |