Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC058167.1 | - | 1 - 2 | 69 | 86 | |
BC080666.1 | - | 81 | 4934956 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |