Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027823.1 | NP_082099.1 | 5 - 6 | 729 | 153 | |
GenBank mRNA | AK082857.1 | - | 5 - 6 | 817 | 152 | |
BC137891.1 | - | 5 - 6 | 729 | 8860910 | 153 | |
BC137892.1 | - | 5 - 6 | 729 | 8860911 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |