Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083917.1 | NP_001077386.1 | 3 - 4 | 631 | 88 | |
NM_001286614.1 | NP_001273543.1 | 3 - 4 | 627 | 88 | ||
NM_178227.4 | NP_839941.1 | 3 - 4 | 631 | 88 | ||
GenBank mRNA | AK049286.1 | - | 3 - 4 | 629 | 88 | |
AK049747.1 | - | 3 - 4 | 450 | 88 | ||
AK076466.1 | - | 2 - 3 | 275 | 88 | ||
AK173115.1 | - | 3 - 4 | 508 | 88 | ||
AY049036.1 | - | 2 - 3 | 236 | 88 | ||
BC032253.1 | - | 3 - 4 | 425 | 5368626 | 88 | |
BC053919.1 | - | 2 - 3 | 526 | 6308278 | 88 | |
BC058083.1 | - | 3 - 4 | 570 | 88 | ||
BC058636.1 | - | 3 - 4 | 630 | 5702879 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |