Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178118.2 | NP_835219.1 | 16 - 17 | 1910 | 86 | |
GenBank mRNA | AK129432.1 | - | 12 - 13 | 1090 | 86 | |
AK142668.1 | - | 10 - 11 | 1034 | 86 | ||
AK147565.1 | - | 10 - 11 | 2650 | 86 | ||
AK147596.1 | - | 10 - 11 | 2650 | 86 | ||
AY549883.1 | - | 16 - 17 | 1768 | 86 | ||
AY549884.1 | - | 16 - 17 | 1771 | 86 | ||
AY549885.1 | - | 12 - 13 | 1060 | 86 | ||
AY549886.1 | - | 12 - 13 | 1069 | 86 | ||
AY549887.1 | - | 10 - 11 | 811 | 86 | ||
BC048182.1 | - | 16 - 17 | 1901 | 6826299 | 86 | |
BC063085.1 | - | 16 - 17 | 1901 | 6826299 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |