Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176860.5 | NP_789830.1 | 5 - 6 | 803 | 89 | |
GenBank mRNA | AB075602.1 | - | 5 - 6 | 798 | 89 | |
AK013361.1 | - | 5 - 6 | 871 | 89 | ||
AK034450.1 | - | 5 - 6 | 994 | 89 | ||
AK035764.1 | - | 5 - 6 | 804 | 89 | ||
AK133948.1 | - | 5 - 6 | 801 | 89 | ||
AK150909.1 | - | 5 - 6 | 1370 | 89 | ||
AK151721.1 | - | 5 - 6 | 1371 | 89 | ||
AK151895.1 | - | 5 - 6 | 1004 | 89 | ||
AK152013.1 | - | 5 - 6 | 1004 | 89 | ||
AK154576.1 | - | 5 - 6 | 1025 | 89 | ||
BC053436.1 | - | 5 - 6 | 775 | 6510702 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |