Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011915.2 | NP_036045.1 | 5 - 6 | 973 | 64 | |
GenBank mRNA | AF122923.1 | - | 5 - 6 | 747 | 64 | |
AK077698.1 | - | 5 - 6 | 973 | 64 | ||
AK142443.1 | - | 5 - 6 | 974 | 64 | ||
AK158359.1 | - | 5 - 6 | 974 | 64 | ||
BC004048.1 | - | 1 - 2 | 253 | 3497155 | 64 | |
BC013268.1 | - | 5 - 6 | 774 | 3984128 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |