Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081193.2 | NP_001074662.2 | 8 - 9 | 2147 | 70 | |
GenBank mRNA | AK140538.1 | - | 8 - 9 | 630 | 70 | |
BC082610.1 | - | 9 - 10 | 911 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |