Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029057.1 | NP_083333.1 | 1 - 2 | 547 | 98 | |
GenBank mRNA | AK044521.1 | - | 1 - 2 | 354 | 98 | |
AK049989.1 | - | 1 - 2 | 379 | 98 | ||
AK173079.1 | - | 1 - 2 | 310 | 98 | ||
BC072607.1 | - | 1 - 2 | 374 | 98 | ||
BC138285.1 | - | 1 - 2 | 754 | 8861304 | 98 | |
BC138286.1 | - | 1 - 2 | 754 | 8861305 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |