Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081056.1 | NP_001074525.1 | 11 - 12 | 1467 | 134 | |
GenBank mRNA | AK029113.1 | - | 6 - 7 | 920 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |