Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172892.3 | NP_766480.1 | 11 - 12 | 1886 | 75 | |
GenBank mRNA | AK036354.1 | - | 11 - 12 | 1888 | 75 | |
AK038937.1 | - | 11 - 12 | 1891 | 75 | ||
BC089161.1 | - | 11 - 12 | 1875 | 30440173 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |