Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033330.3 | NP_001028502.1 | 15 - 16 | 1944 | 91 | |
NM_001290427.1 | NP_001277356.1 | 13 - 14 | 1555 | 91 | ||
NM_001290428.1 | NP_001277357.1 | 12 - 13 | 1488 | 91 | ||
GenBank mRNA | AK147370.1 | - | 15 - 16 | 1945 | 91 | |
AK147440.1 | - | 12 - 13 | 1489 | 91 | ||
BC139429.1 | - | 13 - 14 | 1400 | 8862451 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |