Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037740.1 | NP_001032829.1 | 11 - 12 | 1430 | 76 | |
NM_177204.3 | NP_796178.2 | 11 - 12 | 1430 | 76 | ||
NR_104576.1 | 11 - 12 | 1426 | 76 | |||
GenBank mRNA | AK041004.1 | - | 11 - 12 | 1431 | 76 | |
AK042073.1 | - | 11 - 12 | 1431 | 76 | ||
AK052271.1 | - | 10 - 11 | 1838 | 76 | ||
AK052273.1 | - | 3 - 4 | 455 | 76 | ||
AK122459.1 | - | 11 - 12 | 1235 | 76 | ||
AK163706.1 | - | 11 - 12 | 1430 | 76 | ||
BC147502.1 | - | 11 - 12 | 1422 | 9056385 | 76 | |
BC172034.1 | - | 11 - 12 | 1418 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |