Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291809.1 | NP_001278738.1 | 5 - 6 | 808 | 85 | |
NM_007882.3 | NP_031908.3 | 5 - 6 | 808 | 85 | ||
GenBank mRNA | BC144845.1 | - | 5 - 6 | 793 | 85 | |
Y11169.1 | - | 5 - 6 | 808 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |