Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aoah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281854.1 | NP_001268783.1 | 3 - 4 | 568 | 65 | |
NM_012054.4 | NP_036184.1 | 3 - 4 | 568 | 65 | ||
GenBank mRNA | AK084452.1 | - | 3 - 4 | 563 | 65 | |
AK085343.1 | - | 3 - 4 | 538 | 65 | ||
AK133301.1 | - | 3 - 4 | 509 | 65 | ||
AK150886.1 | - | 3 - 4 | 544 | 65 | ||
AK152346.1 | - | 3 - 4 | 544 | 65 | ||
AK170293.1 | - | 2 - 3 | 104 | 65 | ||
AK171572.1 | - | 3 - 4 | 387 | 65 | ||
BC089580.1 | - | 3 - 4 | 548 | 65 | ||
CT010242.1 | - | 3 - 4 | 287 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |