Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167891.1 | NP_001161363.1 | 5 - 6 | 1219 | 67 | |
GenBank mRNA | AW476657.1 | - | 3 - 4 | 259 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |