Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256382.1 | NP_001243311.1 | 13 - 14 | 2867 | 82 | |
NM_001256383.1 | NP_001243312.1 | 13 - 14 | 2867 | 82 | ||
NM_053271.2 | NP_444501.1 | 15 - 16 | 2987 | 82 | ||
GenBank mRNA | AB021131.1 | - | 15 - 16 | 2941 | 82 | |
AK129206.1 | - | 10 - 11 | 1953 | 82 | ||
BC118979.2 | - | 13 - 14 | 2815 | 40098740 | 82 | |
HM015529.1 | - | 13 - 14 | 2790 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |