Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007883.2 | NP_031909.2 | 7 - 8 | 1017 | 113 | |
GenBank mRNA | AB072269.1 | - | 7 - 8 | 1045 | 113 | |
BC044848.1 | - | 7 - 8 | 978 | 113 | ||
BC131653.1 | - | 7 - 8 | 993 | 40110529 | 113 | |
BC131654.1 | - | 7 - 8 | 993 | 40110531 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |