Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Garem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033445.2 | NP_001028617.2 | 2 - 3 | 627 | 62 | |
GenBank mRNA | AK148318.1 | - | 2 - 3 | 628 | 62 | |
AK220564.1 | - | 2 - 3 | 611 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |