Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081256.1 | NP_001074725.1 | 8 - 9 | 2754 | 88 | |
GenBank mRNA | AK129281.1 | - | 7 - 8 | 2110 | 88 | |
BC060727.1 | - | 1 - 2 | 314 | 88 | ||
BC108415.1 | - | 1 - 2 | 158 | 88 | ||
BC151084.1 | - | 8 - 9 | 2715 | 9087997 | 88 | |
BC172150.1 | - | 7 - 8 | 2115 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |