Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025594.3 | NP_079870.1 | 1 - 2 | 32 | 86 | |
GenBank mRNA | AK012091.1 | - | 1 - 2 | 28 | 86 | |
BC024881.1 | - | 1 - 2 | 32 | 4022326 | 86 | |
BC089501.1 | - | 1 - 2 | 27 | 6594761 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |