Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008214.4 | NP_032240.3 | 8 - 9 | 928 | 93 | |
GenBank mRNA | AK011685.1 | - | 8 - 9 | 844 | 93 | |
AK088824.1 | - | 8 - 9 | 881 | 93 | ||
AK146922.1 | - | 8 - 9 | 881 | 93 | ||
AK150081.1 | - | 8 - 9 | 847 | 93 | ||
AK152329.1 | - | 8 - 9 | 860 | 93 | ||
AK166826.1 | - | 8 - 9 | 844 | 93 | ||
AK166892.1 | - | 8 - 9 | 891 | 93 | ||
BC020088.1 | - | 8 - 9 | 863 | 4008479 | 93 | |
U39473.1 | - | 8 - 9 | 908 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |