Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177615.3 | NP_808283.2 | 8 - 9 | 1345 | 71 | |
GenBank mRNA | AK052641.1 | - | 1284 | 71 | ||
BC089587.1 | - | 8 - 9 | 1282 | 6757541 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |