Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145398.2 | NP_663373.2 | 6 - 7 | 606 | 66 | |
GenBank mRNA | AB091829.1 | - | 6 - 7 | 606 | 66 | |
BC033520.1 | - | 4 - 5 | 230 | 66 | ||
BC125377.1 | - | 6 - 7 | 529 | 40129892 | 66 | |
BC125379.1 | - | 6 - 7 | 529 | 40129894 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |