Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144861.2 | NP_659110.1 | 3 - 4 | 528 | 120 | |
GenBank mRNA | AK137068.1 | - | 3 - 4 | 419 | 120 | |
AK161533.1 | - | 3 - 4 | 383 | 120 | ||
AK220475.1 | - | 3 - 4 | 528 | 120 | ||
BC021395.1 | - | 3 - 4 | 480 | 5067723 | 120 | |
BC023084.1 | - | 3 - 4 | 465 | 4946165 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |