Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153058.4 | NP_694698.3 | 1 - 2 | 249 | 72 | |
GenBank mRNA | AK038562.1 | - | 1 - 2 | 236 | 72 | |
AK040767.1 | - | 1 - 2 | 224 | 72 | ||
AK045493.1 | - | 1 - 2 | 132 | 72 | ||
AK045904.1 | - | 1 - 2 | 224 | 72 | ||
AK046095.1 | - | 1 - 2 | 191 | 72 | ||
AK046221.1 | - | 1 - 2 | 216 | 72 | ||
AK148954.1 | - | 1 - 2 | 224 | 72 | ||
BC025804.1 | - | 1 - 2 | 196 | 5101879 | 72 | |
BC027056.1 | - | 1 - 2 | 215 | 5361238 | 72 | |
BC028987.1 | - | 1 - 2 | 171 | 5362960 | 72 | |
BC035254.1 | - | 1 - 2 | 201 | 5363176 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |