Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130513.1 | NP_001123985.1 | 15 - 16 | 2219 | 92 | |
NM_027286.4 | NP_081562.2 | 14 - 15 | 2071 | 92 | ||
GenBank mRNA | AB053181.1 | - | 14 - 15 | 2075 | 92 | |
AB053182.1 | - | 10 - 11 | 1311 | 92 | ||
AK008530.1 | - | 3 - 4 | 269 | 92 | ||
AK135357.1 | - | 9 - 10 | 1253 | 92 | ||
AK141604.1 | - | 14 - 15 | 1920 | 92 | ||
BC026801.1 | - | 14 - 15 | 2064 | 4236529 | 92 | |
EF408739.1 | - | 15 - 16 | 2180 | 92 | ||
EF408740.1 | - | 15 - 16 | 2180 | 92 | ||
GQ262785.1 | - | 14 - 15 | 1889 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |