Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019776.2 | NP_062750.2 | 18 - 19 | 2325 | 73 | |
GenBank mRNA | AB021491.1 | - | 18 - 19 | 2282 | 73 | |
AK088028.1 | - | 18 - 19 | 2284 | 73 | ||
AK133837.1 | - | 18 - 19 | 2331 | 73 | ||
AK159837.1 | - | 18 - 19 | 2290 | 73 | ||
AK161591.1 | - | 18 - 19 | 2279 | 73 | ||
AK162432.1 | - | 18 - 19 | 2266 | 73 | ||
BC007126.1 | - | 18 - 19 | 2272 | 3592548 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |