Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170745.1 | NP_001164216.1 | 18 - 19 | 3574 | 96 | |
NM_001170746.1 | NP_001164217.1 | 20 - 21 | 3781 | 96 | ||
NM_015823.3 | NP_056638.1 | 19 - 20 | 3616 | 96 | ||
GenBank mRNA | AK039336.1 | - | 16 - 17 | 2601 | 96 | |
AK122349.1 | - | 18 - 19 | 3076 | 96 | ||
AK147530.1 | - | 18 - 19 | 3575 | 96 | ||
BC059005.1 | - | 19 - 20 | 3273 | 6847238 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |