Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029872.3 | NP_001025043.3 | 29 - 30 | 3406 | 60 | |
GenBank mRNA | AK132915.1 | - | 3550 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |