Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138582.1 | NP_613048.1 | 4 - 5 | 818 | 130 | |
GenBank mRNA | AF134319.1 | - | 4 - 5 | 818 | 130 | |
BC137792.1 | - | 4 - 5 | 759 | 8860811 | 130 | |
BC144911.1 | - | 4 - 5 | 759 | 9053459 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |